Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Actual product may vary
Thermo Scientific™

Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays

Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
842040-0650650 mL
842040-0125125 mL
342040-0125125 mL
342040-0250250 mL
342040-0650650 mL
342040-10001 L
842040-10001 L
Catalog number 842040-0650
Price (MXN)
-
Capacity (Metric):
650 mL
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.
Features:
  • Polyethylene terephthalate with natural high density polyethylene closure
  • Molded in graduations and neck ring for use with tamper evident shrink sleeves
  • Gamma irradiated to 10-6 Sterility Assurance Level (SAL)
  • Non-pyrogenic and non-cytotoxic
  • Transparency and reduced permeability to CO2 and O2
  • Bottle and closure material are Animal Derived Component (ADC) Free 
  • Leakproof†

Ordering Information:

Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (English)22 oz.
Certifications/CompliancePET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661>, European Pharmacopeia for Abnormal Toxicity
Closure Size38-430 mm
DescriptionNalgene 650ml PET Square Media Bottle with natural HDPE closure, sterile, made in EU
Diameter (English)2.99 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric)76 mm
Diameter (Metric) Inner Neck28 mm
Height (English)6.8 in.
Height (Metric)173 mm
No. per Case48
No. per Pack24
Product TypeMedia Bottle
ShapeSquare
Capacity (Metric)650 mL
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPET
MouthNarrow
SterilitySterile
Unit SizeEach