Spray Chambers for iCAP™ Series ICP-OES Systems
Thermo Scientific™
Spray Chambers for iCAP™ Series ICP-OES Systems
Thermo Scientific™ Spray Chambers offer improved efficiency and stability of the plasma in standard and specialized ICP-OES applications.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoType
842312051311Baffled Cyclonic Spray Chamber
842312051411Cyclonic Spray Chamber
Número do catálogo 842312051311
Preço (BRL)
-
Type:
Baffled Cyclonic Spray Chamber
Improve efficiency and stability of the plasma in your standard and specialized ICP-OES applications using Thermo Scientific™ Spray Chambers for Thermo Scientific™ iCAP™ Series ICP-OES systems. The single-pass cyclonic spray chamber efficiently filters out larger aerosol droplets whereas the double-pass (baffled) cyclonic spray chamber allows for higher tolerance of dissolved solids and organics.
Improve instrument performance with advanced accessories
These high-purity, quartz spray chambers are suited to ICP-OES applications and compatible with all Thermo Scientific™ 6 mm OD Nebulizers for ICP-OES.
Especificações
For Use With (Equipment)iCAP Series ICP-OES Systems
TypeBaffled Cyclonic Spray Chamber
Unit SizeEach