Product Image
Thermo Scientific™
Liquid Sample Introduction Kits for iCAP™ 6000 Series ICP-OES
Deliver increased flexibility and precise sample introduction from the Thermo Scientific™ iCAP™ 6000 Series ICP-OES.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
842312051791Radial Aqueous Kit
842312051701Radial Organic Kit
842312051711Radial Volatile Organics Kit
842312051731Radial High Solids Kit
842312051721Radial HF Resistant Kit
842312051801Duo Organics Kit
842312051811Duo Volatile Organics Kit
842312051821Duo HF Resistant Kit
842312051831Duo High Solids Kit
842312051891Duo Aqueous Kit
Catalog number 842312051791
Price (TWD)
-
Type:
Radial Aqueous Kit
A range of preconfigured, plug-and-play sample introduction kits offer increased flexibility to analyze a wide variety of sample matrices with the Thermo Scientific™ iCAP™ 6000 Series ICP-OES instruments. With optional, dedicated kits available for high solid content, organic, volatile organic and hydrofluoric acid (HF) containing samples, for the Radial and Duo systems; precise, accurate sample delivery is guaranteed every time.
Increase Flexibility with Sample Introduction Kits
Aqueous
This aqueous sample introduction kit is ideal for liquid sample solutions containing relatively low acid content (except for hydrofluoric acid (HF)) or up to 3% m/v dissolved solids.
The concentric glass nebulizer within this kit is not suitable for solutions containing large suspended particles due to the risk of blockage.
High Solids
In samples such as sea waters, brines and ground waters, geochemical samples and fusion mixtures, with solid particles in suspension (up to 20% m/v dissolved solids), the high solids kit is necessary to prevent blockage problems. Used in conjunction with an argon humidifier, the kit enables extended analysis times without drift.
Organic
This introduction kit is recommended for samples containing relatively nonvolatile, organic solvents, such as xylene, toluene, white spirit and kerosene. These solvents are most likely to be used as diluents for lubricating oils analysis, including additive compounds in virgin oil and wear metals in used oil.
Volatile Organic
Volatile organic solvents, such as alcohols, gasolines and naphthas are often found in petrochemical analysis, both for by-products such as plastics and for petrol itself. The volatile organics sample handling kit is recommended to provide the optimum configuration for analysis by reducing the plasma solvent load.
HF
Un-complexed hydrofluoric acid (HF) is typically obtained during the analysis of geological samples, soils, sediments, ceramics, bricks and cement, together with some types of high temperature alloys. The HF resistant sample handling kit is designed to handle such sample solutions.
Boost Performance with Precision Sample Introduction Kits
Just like our analyzers,Thermo Scientific™ Sample Introduction Kits are designed to maximize convenience, ease of use and precision.
  • All sample-introduction kits are plug and play and self-aligning for convenience in routine maintenance
  • Semi-demountable torch module simply clips into place and automatically interlocks with the system for gas-tight connections
  • Center tube is removable with the torch in situ, for dramatically reduced down time during maintenance and cleaning
Ordering Requirements
Peristaltic pump tubing must be ordered in addition to the sample introduction kits.
Specifications
For Use With (Equipment)iCAP 6000 Series ICP-OES System
TypeRadial Aqueous Kit
Unit SizeEach