Base and Intermediate Tube for Ceramic D-Torches for ICP-OES Systems
Thermo Scientific™
Base and Intermediate Tube for Ceramic D-Torches for ICP-OES Systems
Use dedicated consumables for your Thermo Scientific™ Ceramic D-Torch, to ensure consistent, reliable performance.
Have Questions?
Catalog NumberType
842312052204Base and Intermediate Tube
Catalog number 842312052204
Price (USD)
914.00
Each
Type:
Base and Intermediate Tube
Price (USD)
914.00
Each
Improve ICP-OES plasma torch lifespan by switching to a Thermo Scientific™ Ceramic D-Torch. Use only dedicated consumables for your demountable Ceramic D-Torch, such as this Thermo Scientific™ Base and Intermediate Tube for Ceramic D-Torches to ensure consistent, reliable performance.
Compatibility
Replacement Base and Intermediate Tube compatible with all Thermo Scientific Ceramic D-Torches for ICP-OES systems.
Specifications
For Use With (Equipment)D-Torch for ICP-OES Systems
TypeBase and Intermediate Tube
Unit SizeEach