Internal Standard/Basic Hydride Generation Kit for the iCAP™ Series ICP-OES Systems
Actual product may vary
Thermo Scientific™
Internal Standard/Basic Hydride Generation Kit for the iCAP™ Series ICP-OES Systems
Achieve regulatory compliance for hydride-forming elements in environmental, biological, pharmaceutical and food samples with the Thermo Scientific™ Internal Standards/Basic Hydride Generation Kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
842312052651Mini Pump
842312051551Basic Hydride Kit
Catalog number 842312052651
Price (EUR)
112,00
Each
Type:
Mini Pump
Price (EUR)
112,00
Each
Achieve regulatory compliance for the ICP-OES analysis of toxic hydride-forming elements arsenic, bismuth, antimony, selenium and mercury in environmental, biological, pharmaceutical and food samples. With the simple addition of this Thermo Scientific™ Internal Standards/Basic Hydride Generation Kit for Thermo Scientific™ iCAP™ Series ICP-OES systems, the detection range of the analyzer can be lowered to sub-ppb levels for hydride-forming elements.
Enable regulatory compliance
As maximum allowable levels for these toxic contaminants are driven lower each year, the analytical performance required to achieve the detection limits required by regulations becomes increasingly difficult to achieve using a standard sample introduction setup.
The Basic Hydride Generation Kit sample introduction system provides improved sensitivity for hydride-forming elements.
Specifications
For Use With (Equipment)iCAP 7000/PRO Series ICP-OES Systems
TypeMini Pump
Unit SizeEach