CETAC™ U5000AT+ Ultrasonic Nebulizer
Thermo Scientific™
CETAC™ U5000AT+ Ultrasonic Nebulizer
Increase sensitivity and lower detection limits using this ultrasonic nebulizer for trace element analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
842318051001Ultrasonic Nebulizer220/240 V
842318051002Ultrasonic Nebulizer110/120 V
842318051003Ultrasonic Nebulizer AU220/240 V
Catalog number 842318051001
Price (TWD)
-
Type:
Ultrasonic Nebulizer
Voltage:
220/240 V
Using an ultrasonic nebulizer in conjunction with an optical ICP system is a simple, cost-effective way to increase sensitivity and lower detection limits. While conventional pneumatic nebulizers are generally only 2 to 3% efficient under normal operating conditions, the Thermo Scientific™ CETAC™ U5000AT+ Ultrasonic Nebulizer converts more of the liquid sample into a usable aerosol, with an efficiency of 10 to 15%. The result: enhanced detection limits (up to 10x or greater) for ICP-OES and ICP-MS.
The CETAC U5000AT+ Ultrasonic Nebulizer is compatible with the Thermo Scientific™ iCAP™ Series ICP-OES and Thermo Scientific™ Element™ Series HR-ICP-MS systems.
Lower Detection Limits
  • Up to 10x improvement in analyte signal
  • For ICP-OES detection, limits fall below 1ppb for many elements
  • For ICP-MS, sub-ppt limits can be achieved
  • Even lower limits may be obtained under clean-room conditions
Reliable Unattended Operation
AutoTune Power Supply automatically senses changes in sample composition to ensure uniform and consistent aerosol generation, permitting continuous use of the nebulizer even in the absence of a sample—and reliable unattended operation.
Flexible Options
An optional membrane desolvator (CETAC MDX-200) can be added for further removal of sample solvent (aqueous or volatile organic).
Specifications
Voltage220/240 V
For Use With (Equipment)iCAP ICP-OES System, Element Series HR-ICP-MS Systems
TypeUltrasonic Nebulizer
Unit SizeEach