Product Image
Thermo Scientific™
Sample Loop Assemblies for Aqueous Samples
Designed specifically for aqueous samples, sample loop assemblies are available in a range of volumes to match your analysis requirements.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoTypeDiameter (Metric) InnerVolume (Metric)
842347004050Aqueous Sample Loop2 mm2.0 mL
842347004048Aqueous Sample Loop2 mm1.0 mL
842347004051Aqueous Sample Loop2 mm3.0 mL
842347004053Aqueous Sample Loop2 mm5.0 mL
842347004047Aqueous Sample Loop2 mm0.7 mL
842347004049Aqueous Sample Loop2 mm1.5 mL
842347004052Aqueous Sample Loop2 mm4.0 mL
Número do catálogo 842347004050
Preço (BRL)
-
Type:
Aqueous Sample Loop
Diameter (Metric) Inner:
2 mm
Volume (Metric):
2.0 mL
Customize your consumables to match your chosen sample matrix precisely to maximize efficiency and precision. These Thermo Scientific™ Sample Loop Assemblies for Aqueous Samples are available in a range of volumes to match your analysis requirements.
Choose Customized Consumables to Suit your Analysis
Sample loop assembly aqueous 2mm ID, 0.7ml
Sample loop assembly aqueous 2mm ID, 1.0ml
Sample loop assembly aqueous 2mm ID, 1.5ml
Sample loop assembly aqueous 2mm ID, 2.0ml
Sample loop assembly aqueous 2mm ID, 3.0ml
Sample loop assembly aqueous 2mm ID, 4.0ml
Sample loop assembly aqueous 2mm ID, 5.0ml
Especificações
Diameter (Metric) Inner2 mm
Volume (Metric)2.0 mL
For Use With (Equipment)Mass Spectrometers
TypeAqueous Sample Loop
Unit SizeEach