Pierce™ Streptavidin, DyLight™ 650 Conjugated

Catalog number: 84547B

Thermo Scientific™  Related applications: Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Streptavidin, DyLight 650 Conjugate are preparations of streptavidin biotin-binding protein that are tagged with Thermo Scientific DyLight Fluorophores for use in high-performance fluorescence assays and cellular imaging.

Features of Streptavidin, DyLight 650 Conjugate:

Streptavidin protein—choose the biotin-binding protein that is best for your application
Optimized conjugates—manufactured with optimal dye:protein ratios to provide the highest possible binding function and overall fluorescence intensity
Consistent and stable—all conjugates are stable for 1 year at 4°C, providing consistent performance for the duration of use

Features of DyLight Fluorophore:

Bright fluorescence—intense emission provides superior sensitivity and requires less conjugate
Narrow emission spectra—negligible bleed-through between fluorophore channels enables multi-color detection
Excellent photo-stability—exceptional resistance to photo-bleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4Pi microscopy)
Buffer stability—conjugated fluorophores are completely stable at pH 4-9
Instrument compatibility—excitation and emission spectra correspond with filter sets and laser settings of all popular fluorescence instrumentation

Streptavidin biotin-binding protein is offered as highly functional conjugate with each of nine different DyLight Fluorescent Dyes having absorption spectra ranging from 350nm to 777nm and bright emission spectra that match the principal output wavelengths of common fluorescence microscopes and imagers. DyLight Conjugates exhibit higher fluorescence intensity and photostability than Alexa Fluor*, Cy* and LI-COR* Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9).

Related Products
Pierce™ Streptavidin, DyLight™ 405 Conjugated
Pierce™ Streptavidin, DyLight™ 488 Conjugated
Pierce™ Streptavidin, DyLight™ 550 Conjugated
Pierce™ Streptavidin, DyLight™ 594 Conjugated
Pierce™ Streptavidin, DyLight™ 633 Conjugated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: DyLight™ 650
Product Line: DyLight™, Pierce™
Product Size: Custom
Target Molecule: Biotin

Contents & storage

Stable for 1 year at 4°C

Documents

Manuals & protocols