Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »
Power Cord with C/13 Connector, United Kingdom
Catalog number: 84860
Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This electrical power cord has a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets used in the United Kingdom.
This black, 2.4- to 3.5m-long, cord has been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).
Related Products
myECL™ Imager
Pierce™ Power Station
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): Pierce™ Power System, myECL™ Imager
Product Size: 1 cord
Type: Cords and Connectors
Contents & storage
0.0
Documents
Manuals & protocols