Can I purchase the Reversed-Phase Fractionation Spin Columns from the Pierce High pH Reversed-Phase Peptide Fractionation Kit (Cat. No. 84868) separately?
No, the spin columns from this kit cannot be purchased separately. However, if you are only using these columns to remove salts from your peptide samples, you can purchase the Pierce Peptide Desalting Spin Columns (Cat. No. 89851, 89852), as an alternative.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.