SPoT-Light® CISH Polymer Detection Kit

Catalog number: 849246

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This CISH detection kit will detect digoxigenin (DIG)-labeled probes. It can be used only for CISH. Since the kit uses a polymerized HRP⁄DAB detection system to localize DIG labeled probes, the signal of CISH staining is up to 50% stronger than the previous CISH Detection Kit (Cat No. 84-9243). The kit is optimized for SPOT-Light probes to detect gene amplification and gene deletion.

Applications: CISH (Chromogenic In Situ Hybridization)
For In Vitro Diagnostic Use.

Specifications

Substrate: DAB
Label or Dye: HRP (Horseradish Peroxidase), Digoxigenin
Product Size: 1 kit
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Molecule: DNA
Detection Method: Colorimetric
Target Specificity: Not Target-Specific
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products