HistoMouse-Plus Kit, AEC, broad spectrum

Catalog number: 859541

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The HistoMouse-Plus kit is the 2nd generation HistoMouse kit for the detection of mouse primary antibodies on mouse tissue samples without causing background due to endogenous mouse Ig. Our patented BEAT™ technology (Blocking Endogenous Antibody Technology) is used to prevent the secondary antibody conjugate from reacting with endogenous mouse Ig. The LAB-SA detection method is used to amplify the signal. In addition, our HistoMouse-Plus kit includes the AEC Single Solution chromogen⁄ substrate system to create an intense red deposit around the antigen⁄antibody⁄enzyme complex in the sample.

Reactivity: Mouse, Rabbit, Rat, G. Pig

Applications: Immunohistochemistry (IHC)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Colorimetric
For Use With (Equipment): Light Microscope
Product Line: HistoMouse™
Product Size: 50 slides
Reactivity: Guinea Pig, Mouse, Rabbit, Rat
Substrate: AEC
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Mouse
Target Specificity: Not Target-Specific
Technique: HistoMouse
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products