Histostain-Plus IHC Kit, AEC, broad spectrum

Catalog number: 859943

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Full Product Description
Histostain-Plus kits use 2nd generation LAB-SA technology and are ideal for general immunohistochemical applications. Histostain-Plus kits offer superior sensitivity without compromising background. They are designed to detect mouse, rabbit, rat, or guinea pig primary antibodies that are bound to human tissue or cell antigens. Histostain-Plus kits are excellent for frozen and formalin-fixed, paraffin-embedded tissue sections, cell smears, etc. These kits are very easy to use: all components are liquid (except Fast-Red) and come in dropper bottles. No reconstitution, no pipetting, no measuring is necessary, and the enzyme is conjugated to streptavidin to eliminate the preincubation complexing step. DAB chromogen yields a brown stain, AEC yields a deep red stain, and Fast-Red yields a bright red stain.

Reactivity: Mouse, Rabbit, Rat, Guinea Pig

Applications: Immunohistochemistry (IHC)
For In Vitro Diagnostic Use.


Substrate: AEC
Technique: LAB-SA (Labeled Streptavidin-Biotin)
Reactivity: Mouse, Guinea Pig, Rabbit, Rat
Product Size: 150 slides
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Human
Detection Method: Colorimetric
Target Specificity: Not Target-Specific
For Use With (Equipment): Light Microscope
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).


Manuals & protocols

Recommended products