Thermo Scientific™
Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Periodic Acid)
Related applications:
Anatomical Pathology
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Thermo Scientific™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution.
Back to top
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green