Thermo Scientific™
Richard-Allan Scientific™ Steiner-Steiner Silver
Related applications:
Anatomical Pathology
Determine the presence of spirochetes and nonfilamentous bacteria with Thermo Scientific™ Richard-Allan Scientific™ Steiner-Steiner Silver.
Back to top
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results