96-Well Flipper™ Microtube Racks
Thermo Scientific™
96-Well Flipper™ Microtube Racks
Protect samples and allow racks to be stacked using this 96-well microtube rack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorUnit Size
8780Fluorescent GreenCase of 10
8760Fluorescent PinkCase of 10
8760PK
also known as 8760
Fluorescent PinkEach
8790PK
also known as 8790
RedPack of 1
Catalog number 8780
Price (USD)
-
Color:
Fluorescent Green
Unit Size:
Case of 10
Protect samples and allow racks to be stacked using the Thermo Scientific™ 96-Well Flipper™ Microtube Rack. The rack, which is molded with an alphanumeric grid for easy tube identification, holds 0.5mL tubes on one side and 1.5mL tubes on the other.
  • Holds 0.5mL tubes on one side, 1.5mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Sides have a frosted writing surface for smear-resistant labeling
  • Several colors available
Specifications
DescriptionFlipper Rack
No. per Case10 Racks
ColorFluorescent Green
HoldsMicrotubes (2 mL), Microtubes (1.5 mL), Microtubes (2 mL)
MaterialPolypropylene
Product Type96-well Flipper Rack
ShapeRectangular
StackableYes
Unit SizeCase of 10