HistoMouse-MAX Kit, AEC, broad spectrum

Catalog number: 879541

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This HistoMouse™-MAX kit is designed to allow immunohistochemical studies of mouse or rat tissue samples using mouse, rabbit, rat and guinea pig primary antibodies. The kit incorporates a highly sensitive horseradish peroxidase (HRP) second antibody to increase sensitivity. HistoMouse™ MAX detection system is faster and cleaner because it uses one step instead of two steps, as in the LAB-SA method for the secondary antibody step. After the user-supplied primary antibody is incubated with the sample, an HRP-conjugated, affinity-purified secondary antibody is bound to the primary antibody . A chromogen/substrate system then creates an intense deposit around the antigen/antibody/enzyme complex in the sample.

Reactivity: Mouse, Rabbit, Rat, G. Pig

Applications: Immunohistochemistry
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Colorimetric
For Use With (Equipment): Light Microscope
Product Line: HistoMouse™-MAX
Product Size: 50 slides
Reactivity: Guinea Pig, Mouse, Rabbit, Rat
Substrate: AEC
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Mouse, Rat
Target Specificity: Not Target-Specific
Technique: HistoMouse
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols