Notice: This product will be discontinued once remaining inventory is depleted.
IL-21 Mouse ELISPOT Kit
Invitrogen™

IL-21 Mouse ELISPOT Kit

Have Questions?
Catalog NumberQuantity
88-7210-8810 x 96 Tests
Catalog number 88-7210-88
Price (USD)
-
Quantity:
10 x 96 Tests
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by NKT, T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl-6. As with IFN gamma for Th1, IL-4 for Th2 cells, and IL-17A for Th17 cells, IL-21 is critical for TFH effector function. This cytokine plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naïve B cells.

In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. High levels of IL-21 are present in chemically-induced colitis models. IL-21-deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules.

Reported Application
ELISPOT

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeSee product documentation
Assay SensitivitySee product documentation
ConjugateBiotin
For Use With (Equipment)Microplate Reader
Former IdentifiersReady-SET-Go!, eBioscience
Geneinterleukin 21
Gene AliasIL-21
Gene ID (Entrez)60505
Gene SymbolIL21
Label or DyeHRP
ProteinInterleukin-21
Protein AliasesIL-21
Quantity10 x 96 Tests
Target Kit NamedIL-21
Target Species ValidatedMouse
UniProt IDQ9ES17
Detection MethodColorimetric
Immunoassay Kit FormatELISPOT Kit
Target SpeciesMouse
Unit Size10 x 96 tests
Contents & Storage
Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer: This ELISPOT Kit may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 μM).
Assay Diluent: 5X Concentrated
Detection Enzyme: Pre-titrated Avidin-HRP
Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and enzyme