Thermo Scientific™
Richard-Allan Scientific™ Elastic Stain
Related applications:
Anatomical Pathology
Demonstrate the atrophy, thinning and loss of elastic tissue with Thermo Scientific™ Richard-Allan Scientific™ Elastic Stain.
Back to top
  • Based on the Verhoeff Elastic technique
  • Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
  • Elastic fibers are stained blue-black to-black with red collagen and a yellow background