Thermo Scientific™
Richard-Allan Scientific™ Iron Stain
 Related applications:
Anatomical Pathology
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Thermo Scientific™ Richard-Allan Scientific™ Iron Stain.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
  • Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
Documents
Manuals & protocols