Thermo Scientific™
Richard-Allan Scientific™ Periodic Acid-Schiff (PAS)
Related applications:
Anatomical Pathology
Demonstrate the patterns and presence of specific carbohydrates with Thermo Scientific™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution
Back to top
  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent