Product Image
Invitrogen™

MagniSort™ Human CD4 T cell 2-Step Enrichment Kit

The MagniSort™ Human CD4 2-Step Enrichment Kit is designed for the magnetic separation of CD4+ T cells by positive selection深入閱讀
Have Questions?
產品號碼Quantity
8802-6831-74200 tests
產品號碼 8802-6831-74
價格 (TWD)
***
Quantity:
200 tests
價格 (TWD)
***

The MagniSort™ Human CD4 2-Step Enrichment Kit is designed for the magnetic separation of CD4+ T cells by positive selection after first depleting monocytes. It has been optimized for the isolation of CD4+ T cells from human peripheral blood monocuclear cells, utilizing biotinylated Anti-Human CD14 and Anti-Human CD4 antibodies, as well as streptavidin-coated magnetic beads. In the first step, monocytes are bound by Anti-Human CD14 antibody and then magnetic beads. When placed in a magnetic field, the monocytes are held in the magnet while the remaining cells are decanted and used for the second step. In the second step, the monocyte-depleted cells are bound by Anti-Human CD4 antibody and then magnetic beads. Now, when placed in the magnetic field, the CD4+ T cells are held in the magnet while undesired cells can be decanted.

After enrichment, the purity of selected cells can be verified by staining with Anti-Human CD4, clones OKT4 or SK3.

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
規格
Cell TypeCD4+
FormatKit
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
For Use With (Application)Magnetic Cell Separation
Product TypeCell Enrichment Kit
Unit SizeEach