Product Image
Invitrogen™
MagniSort™ Mouse CD45.2 Positive Selection Kit
The MagniSort™ Mouse CD45.2 Positive Selection Kit is designed for the magnetic separation of CD45.2+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6849-74200 tests
Catalog number 8802-6849-74
Price (KRW)
930,000
Online offer
Ends: 30-Sep-2026
1,094,000
Save 164,000 (15%)
Each
Quantity:
200 tests
Price (KRW)
930,000
Online offer
Ends: 30-Sep-2026
1,094,000
Save 164,000 (15%)
Each
The MagniSort™ Mouse CD45.2 Positive Selection Kit is designed for the magnetic separation of CD45.2+ cells by positive selection. It has been optimized for the isolation of CD45.2+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD45.2 antibody and streptavidin-coated magnetic beads. CD45.2+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD45.2+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD45.2, clone 104.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD45.2+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach