Thermo Scientific™
Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Periodic Acid)
 Related applications:
Anatomical Pathology
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Thermo Scientific™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Documents
Manuals & protocols