Thermo Scientific™
Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen)
 Related applications:
Anatomical Pathology
Save time with Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
  • Stains collagen blue, cytoplasm and muscle fibers red and the nuclei blue-to-black
Documents
Manuals & protocols