Thermo Scientific™
Richard-Allan Scientific™ Mucicarmine
Related applications:
Anatomical Pathology
Identify primary tumor sites with the aid of Thermo Scientific™ Richard-Allan Scientific™ Mucicarmine
Back to top
  • Aids in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
  • Mucin is stained dark rose and the nuclei are black
  • Capsule of Cryptococcus (fungus) is stained deep rose
  • Tartrazine solution is used as a yellow cytoplasmic stain