Product Image
Invitrogen™
MagniSort™ Human CD45 Depletion Kit
The MagniSort™ Human CD45 Depletion Kit is designed to deplete CD45+ cells from human lysed whole blood or peripheral bloodRead more
Have Questions?
Catalog NumberQuantity
8804-6802-74200 tests
Catalog number 8804-6802-74
Price (HKD)
7,001.00
Online exclusive
Ends: 31-Dec-2026
8,262.00
Save 1,261.00
Each
Quantity:
200 tests
Price (HKD)
7,001.00
Online exclusive
Ends: 31-Dec-2026
8,262.00
Save 1,261.00
Each
The MagniSort™ Human CD45 Depletion Kit is designed to deplete CD45+ cells from human lysed whole blood or peripheral blood mononuclear cells, utilizing a biotinylated Anti-Human CD45 antibody and streptavidin-coated magnetic beads. CD45+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting.
After depletion, the purity of selected cells can be verified by staining with Anti-Human CD45, clone 2D1.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD45+
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeCell Depletion Kit
Unit SizeEach