Product Image
Invitrogen™

MagniSort™ Human CD4 Memory T cell Enrichment Kit

The MagniSort™ Human CD4 Memory T cell Enrichment Kit is designed for the magnetic separation of CD4 memory T cells by negative selection.
Have Questions?
Catalog NumberQuantity
8804-6813-74200 tests
Catalog number 8804-6813-74
Price (CLP)
1.206.534
Each
Quantity:
200 tests
Price (CLP)
1.206.534
Each

The MagniSort™ Human CD4 Memory T cell Enrichment Kit is designed for the magnetic separation of CD4 memory T cells by negative selection. It has been optimized for the isolation of human CD4 memory T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 memory T cells untouched and free in solution.

The MagniSort™ Human CD4 Memory T cell Enrichment Antibody Cocktail contains the following antibodies:

  • Anti-Human CD8 Biotin
  • Anti-Human CD11b Biotin
  • Anti-Human CD14 Biotin
  • Anti-Human CD16 Biotin
  • Anti-Human CD19 Biotin
  • Anti-Human CD20 Biotin
  • Anti-Human CD36 Biotin
  • Anti-Human CD45RA Biotin
  • Anti-Human CD56 Biotin
  • Anti-Human CD123 Biotin
  • Anti-Human CD235a Biotin
  • Anti-Human γδ TCR Biotin

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (Memory, CD4)
FormatKit
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
For Use With (Application)Magnetic Cell Separation
Product TypeCell Enrichment Kit
Unit SizeEach