Product Image
Invitrogen™
MagniSort™ Mouse T cell Enrichment Kit
The MagniSort™ Mouse T cell Enrichment Kit is designed for the magnetic separation of mouse T cells by negative selection.
Have Questions?
Catalog NumberQuantity
8804-6820-74200 tests
Catalog number 8804-6820-74
Price (EUR)
599,65
Online exclusive
800,00
Save 200,35 (25%)
Each
Quantity:
200 tests
Price (EUR)
599,65
Online exclusive
800,00
Save 200,35 (25%)
Each
The MagniSort™ Mouse T cell Enrichment Kit is designed for the magnetic separation of mouse T cells by negative selection. It has been optimized for the isolation of mouse T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave T cells untouched and free in solution.
The MagniSort™ Mouse T cell Enrichment Antibody Cocktail contains the following antibodies:
  • Anti-Mouse CD11b Biotin
  • Anti-Mouse CD19 Biotin
  • Anti-Mouse CD24 Biotin
  • Anti-Mouse CD45R (B220) Biotin
  • Anti-Mouse CD49b (Integrin alpha 2) Biotin
  • Anti-Mouse Ly-6G (Gr-1) Biotin
  • Anti-Mouse TER-119 Biotin
Reported Application: Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Enrichment Kit
Unit SizeEach