Product Image
Invitrogen™

MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cellsRead more
Have Questions?
Catalog NumberQuantity
8804-6825-74200 tests
Catalog number 8804-6825-74
Price (CLP)
1.094.194
Each
Quantity:
200 tests
Price (CLP)
1.094.194
Each

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of mouse CD8 naïve T cells from spleens or lymphnodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:

  • Anti-Mouse CD4 Biotin
  • Anti-Mouse CD11b Biotin
  • Anti-Mouse CD19 Biotin
  • Anti-Mouse CD24 Biotin
  • Anti-Mouse CD44 Biotin
  • Anti-Mouse CD45R (B220) Biotin
  • Anti-Mouse CD49b (Integrin alpha 2) Biotin
  • Anti-Mouse Ly-6G (Gr-1) Biotin
  • Anti-Mouse γδ TCR Biotin
  • Anti-Mouse TER-119 Biotin

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (Naïve, CD8+)
FormatKit
No. of Tests200 Tests
Quantity200 tests
Target SpeciesMouse
For Use With (Application)Magnetic Cell Separation
Product TypeCell Enrichment Kit
Unit SizeEach