Product Image
Invitrogen™
MagniSort™ Human Hematopoietic Lineage Depletion Kit
The MagniSort™ Human Hematopoietic Lineage Depletion Kit is designed for the magnetic depletion of lineage positive cells from normal humanRead more
Have Questions?
Catalog NumberQuantity
8804-6836-74200 tests
Catalog number 8804-6836-74
Price (USD)
-
Quantity:
200 tests
The MagniSort™ Human Hematopoietic Lineage Depletion Kit is designed for the magnetic depletion of lineage positive cells from normal human peripheral blood mononuclear cells. The kit utilizes a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells, including T cells, B cells, NK cells, myeloid cells, granulocytes, and red blood cells, are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave lineage negative cells untouched and free in solution.
The MagniSort™ Human Hematopoietic Lineage Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD10 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypePBMCs
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeCell Depletion Kit
Unit SizeEach