Product Image
Invitrogen™
MagniSort™ Human pan-Monocyte Enrichment Kit
The MagniSort™ Human Pan-Monocyte Enrichment Kit is designed for the magnetic separation of conventional CD14+ and unconventional CD16+CD14- monocytes byRead more
Have Questions?
Catalog NumberQuantity
8804-6837-74200 tests
Catalog number 8804-6837-74
Price (KRW)
1,112,000
Online offer
Ends: 31-Dec-2026
1,308,000
Save 196,000 (15%)
Each
Quantity:
200 tests
Price (KRW)
1,112,000
Online offer
Ends: 31-Dec-2026
1,308,000
Save 196,000 (15%)
Each
The MagniSort™ Human Pan-Monocyte Enrichment Kit is designed for the magnetic separation of conventional CD14+ and unconventional CD16+CD14- monocytes by negative selection. It has been optimized for the isolation of human pan-monocytes from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells, including CD16+ NK cells, are bound by antibody and then magnetic beads that, when placed in a magnetic field, leaving monocytes untouched and free in solution.
The MagniSort™ Human Pan-Monocyte Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD7 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD56 Biotin
Anti-Human CD57 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Human Fc Receptor Binding Inhibitor Purified
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMonocytes (pan)
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeHuman Pan-Monocyte Enrichment Kit
Unit SizeEach