Product Image
Invitrogen™
MagniSort™ Mouse CD45 Depletion Kit
The MagniSort™ Mouse CD45 Depletion Kit is designed for the magnetic depletion of mouse CD45+ cells from mouse spleens, boneRead more
Have Questions?
Catalog NumberQuantity
8804-6864-74200 tests
Catalog number 8804-6864-74
Price (KRW)
-
Quantity:
200 tests
The MagniSort™ Mouse CD45 Depletion Kit is designed for the magnetic depletion of mouse CD45+ cells from mouse spleens, bone marrow or digested lung tissue. It utilizes a biotinylated Anti-Mouse CD45 antibody and streptavidin-coated magnetic beads. CD45+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting.
After depletion, the purity of enriched CD45- cells can be verified by staining with Anti-Mouse CD45, clone 30-F11.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD45+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Depletion Kit
Unit SizeEach