Product Image
Invitrogen™

MagniSort™ Human B Cell Enrichment Kit (2nd Generation)

The MagniSort™ Human B cell Enrichment Kit (2nd Generation) is designed for the magnetic separation of B cells by negative selection with improved purity.
Have Questions?
Catalog NumberQuantity
8804-6867-74200 tests
Catalog number 8804-6867-74
Price (EUR)
709,84
Online exclusive
934,00
Save 224,16 (24%)
Each
Quantity:
200 tests
Price (EUR)
709,84
Online exclusive
934,00
Save 224,16 (24%)
Each

The MagniSort™ Human B cell Enrichment Kit (2nd Generation) is designed for the magnetic separation of B cells by negative selection with improved purity. It has been optimized for the isolation of human B cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave B cells untouched and free in solution.

The MagniSort™ Human B cell Enrichment Antibody Cocktail (2nd Generation) contains the following antibodies:

  • Anti-Human CD2 Biotin
  • Anti-Human CD3 Biotin
  • Anti-Human CD11b Biotin
  • Anti-Human CD14 Biotin
  • Anti-Human CD16 Biotin
  • Anti-Human CD36 Biotin
  • Anti-Human CD56 Biotin
  • Anti-Human CD235a Biotin

Reported Application: Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeB Cells
FormatKit
No. of Tests200 Tests
Quantity200 tests
Target SpeciesHuman
For Use With (Application)Magnetic Cell Separation
Product TypeCell Enrichment Kit
Unit SizeEach