Can I also stain for intracellular targets such as transcription factors and cytokines using the eBioscience BrdU staining kit?
Yes, primary antibodies against additional intracellular targets can be added alongside the Anti-BrdU antibody. However, it is critical that you empirically determine whether these additional antibodies are compatible with this staining protocol.