Which 96-well plate do you recommend using with the Pierce LAL Chromogenic Endotoxin Quantitation Kit (Cat. No. 88282)?
Any endotoxin-free (EF) 96-well plate can be used with the Pierce LAL Chromogenic Endotoxin Quantitation Kit. Please check the below EF products:
- EF microplates (https://www.fishersci.com/shop/products/endotoxin-free-96wl-mcro-50-cs/NC9217517)
- Thermo Scientific Snap Cap Low Retention Microcentrifuge Tubes (https://www.thermofisher.com/order/catalog/product/3453)
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
- EF microplates (https://www.fishersci.com/shop/products/endotoxin-free-96wl-mcro-50-cs/NC9217517)
- Thermo Scientific Snap Cap Low Retention Microcentrifuge Tubes (https://www.thermofisher.com/order/catalog/product/3453)
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.