Detergent Removal Spin Columns & Plates
Thermo Scientific™

Detergent Removal Spin Columns & Plates

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct LineProduct Type
883072 x 96-well PlatesHiPPRSpin Plate
8778010 mLPierceDetergent Removal Resin
8777625 x 125 μL ColumnsPierceSpin Column
8777725 x 500 μL ColumnsPierceSpin Column
877785 x 2 mL ColumnsPierceSpin Column
877795 x 4 mL ColumnsPierceSpin Column
883042 x 96-well PlatesPierceSpin Plate
883055 mLHiPPRSpin Column
8830624 x 0.1 mL ColumnsHiPPRSpin Column
Catalog number 88307
Price (USD)
622.65
Online exclusive
684.00
Save 61.35
Each
Quantity:
2 x 96-well Plates
Product Line:
HiPPR
Product Type:
Spin Plate
Price (USD)
622.65
Online exclusive
684.00
Save 61.35
Each

Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.

  • Optimized—removes >95% of detergent from samples of proteins or peptides
  • Fast—sample processing takes less than 15 minutes
  • Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin

Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit

HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:

  • Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
  • Spin columns—0.1 mL micro-centrifuge columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeProprietary Resin, Affinity
Description96-well Filter Plate
Purification TargetDetergent Removal
Quantity2 x 96-well Plates
Stationary PhaseProprietary Ligand
Sufficient For192 Samples
Product LineHiPPR
Product TypeSpin Plate
Unit SizeEach
Contents & Storage
• HiPPR Detergent Removal 96-well Filter Spin Plate, 2 plates
• 96-well Deep-well collection plate, 2 plates
• 96-well collection plate, 2 plates

Store at 4°C.