DMEM for SILAC, 500 mL - FAQs
DMEM for SILAC, 500 mL - FAQs
View additional product information for DMEM for SILAC - FAQs (A33822, 88364)
1 product FAQs found
Does DMEM for SILAC (Cat. No. 88364, A33822) have the labeled amino acids in it or do those have to be added separately?
DMEM for SILAC (Cat. No. 88364, A33822) is a liquid medium deficient in both L-lysine and L-arginine. It is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.