eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 615
Product Image
Invitrogen™

eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 615

This BrdU Kit for immunohistochemistry (IHC)/immunocytochemistry (ICC) with immunofluorescence detection contains the necessary reagents and buffers for identifying and examining자세히 알아보기
Have Questions?
카탈로그 번호수량
8842-6599-4550 tests
카탈로그 번호 8842-6599-45
제품 가격(KRW)
630,000
온라인 행사
Ends: 31-Mar-2026
741,000
할인액 111,000 (15%)
Each
카트에 추가하기
수량:
50 tests
제품 가격(KRW)
630,000
온라인 행사
Ends: 31-Mar-2026
741,000
할인액 111,000 (15%)
Each
카트에 추가하기
This BrdU Kit for immunohistochemistry (IHC)/immunocytochemistry (ICC) with immunofluorescence detection contains the necessary reagents and buffers for identifying and examining proliferating cells by immunohistochemical or immunocytochemical analysis. Cells are labeled in vitro or in vivo with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine, which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Following fixation and antigen retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using a streptavidin-conjugated fluorophore. This kit has been optimized for IHC with both frozen and formalin-fixed paraffin embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.

Conjugate
eFluor 615

Emit
615 nm

Excite
595 nm

Reported Application
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Microscopy, Immunocytochemistry

For Research Use Only. Not for use in diagnostic procedures.
사양
검출 방법Fluorometric
용도(애플리케이션)ICC (Immunochemistry), IHC (Immunohistochemistry)
제품 유형IHC/ICC BrdU Kit
수량50 tests
타겟 종All species
콘주게이트BrdU
제품라인eBioscience
Unit SizeEach