Product Image
Invitrogen™

eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 660

This BrdU Kit for immunohistochemistry (IHC)/immunocytochemistry (ICC) with immunofluorescence detection contains the necessary reagents and buffers for identifying and examiningLeia mais
Have Questions?
Número do catálogoQuantity
8850-6599-4550 tests
Número do catálogo 8850-6599-45
Preço (BRL)
4.422,21
Each
Quantity:
50 tests
Preço (BRL)
4.422,21
Each
This BrdU Kit for immunohistochemistry (IHC)/immunocytochemistry (ICC) with immunofluorescence detection contains the necessary reagents and buffers for identifying and examining proliferating cells by immunohistochemical or immunocytochemical analysis. Cells are labeled in vitro or in vivo with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine, which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Following fixation and antigen retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using a streptavidin-conjugated fluorophore. This kit has been optimized for IHC with both frozen and formalin-fixed paraffin embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.

Conjugate
eFluor 660

Laser
Red Laser

Emit
668 nm

Excite
633 - 647 nm

Reported Application
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry, Microscopy

For Research Use Only. Not for use in diagnostic procedures.
Especificações
Detection MethodFluorometric
For Use With (Application)ICC (Immunochemistry), IHC (Immunohistochemistry)
Product TypeIHC/ICC BrdU Kit
Quantity50 tests
Target SpeciesAll species
ConjugateBrdU
Product LineeBioscience
Unit SizeEach