Pierce™ NHS-Activated Magnetic Beads
Thermo Scientific™

Pierce™ NHS-Activated Magnetic Beads

Immobilize proteins or other ligands for magnetic separation affinity purification with these magnetic particles coated with amine-reactive NHS-ester groups.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
888261 mL
888275 mL
카탈로그 번호 88826
제품 가격(KRW)
431,000
온라인 행사
Ends: 31-Dec-2026
492,000
할인액 61,000
Each
수량:
1 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
431,000
온라인 행사
Ends: 31-Dec-2026
492,000
할인액 61,000
Each

Thermo Scientific Pierce NHS-Activated Magnetic Beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.

Features of NHS-Activated Magnetic Beads:

  • High capacity—at least four times higher binding capacity than NHS-activated magnetic beads from other suppliers
  • Easy to use—immobilize in a simple one-step reaction with minimal hands-on time
  • Safe—no hazardous chemicals needed (e.g., sodium cyanoborohydride and cyanogen bromide)
  • Ligand-compatible—use to immobilize with nearly any primary amine-containing compound or affinity ligand
  • Low non-specific binding—the bead surface is pre-blocked and any nonreacted NHS-ester groups are fully quenched
  • Protocol-compatible—protein coupling to the beads and downstream applications can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)

The activated magnetic beads contain N-hydroxy-succinimide (NHS) functional groups that react with primary amines forming stable amide linkages. Once they are covalently attached, the immobilized proteins are highly resistant to leaching from the bead surface. When prepared beads are used in experiments, nonspecific binding is negligible because nonreacted NHS-ester groups are thoroughly blocked during the coupling procedure. Pierce NHS-Activated Magnetic beads can be coupled and processed either manually with a magnetic stand or with automated platforms such as the Thermo Scientific KingFisher Instruments.

Applications:

  • Immobilization of ligands for the purification of recombinant proteins
  • Immobilization of antibodies for immunoprecipitations and co-immunoprecipitations free of antibody contamination in the eluates
  • Immobilization of secondary antibody for the affinity purification of antibody subtypes from ascites, serum and cell culture supernatant

Pierce NHS-Activated Magnetic Beads offer a convenient way to conjugate any desired protein to a magnetic bead surface. The process does not require hazardous chemicals or lengthy reaction schemes. The beads are first incubated with protein for 1 to 2 hours in an amine free buffer at pH 7 to 9 to allow for covalent coupling through NHS-ester chemistry. The beads are subsequently washed and then any remaining active NHS-ester groups are quenched. The amine reactive chemistry and subsequent quench are completed in 3 to 4 hours. Coupled protein does not leach from the bead surface. In addition, the beads exhibit very low non-specific binding due to effective quenching and a proprietary blocking agent that coats the surface of the base particle. Following conjugation the prepared magnetic beads are typically used in affinity purification procedures.

For Research Use Only. Not for use in diagnostic procedures.
Pierce NHS-Activated Magnetic Beads offer a convenient way to conjugate any desired protein to a magnetic bead surface. The process does not require hazardous chemicals or lengthy reaction schemes. The beads are first incubated with protein for 1 to 2 hours in an amine free buffer at pH 7 to 9 to allow for covalent coupling through NHS-ester chemistry. The beads are subsequently washed and then any remaining active NHS-ester groups are quenched. The amine reactive chemistry and subsequent quench are completed in 3 to 4 hours. Coupled protein does not leach from the bead surface. In addition, the beads exhibit very low non-specific binding due to effective quenching and a proprietary blocking agent that coats the surface of the base particle. Following conjugation the prepared magnetic beads are typically used in affinity purification procedures.
사양
용량(미터법)1 mL
농도Slurry: 10 mg/mL, 1% solids
단백질 형태Recombinant
수량1 mL
표면 기능성NHS Ester
타겟Amine
형태Liquid
입자 크기1 μm
제품라인Pierce
유형Magnetic Affinity Bead
Unit SizeEach
구성 및 보관
Upon receipt store at 4°C. Product shipped with an ice pack.

자주 묻는 질문(FAQ)

Can you provide the shelf-life for NHS-Activated Magnetic Beads?

NHS-Activated Magnetic Beads is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What should I do to increase the coupling efficiency of antibodies to the Pierce NHS-Activated Magnetic Beads?

-Make sure all primary amine-containing buffer components of the buffer like Tris or glycine are completely removed by dialysis, desalting, or the use of the Pierce Antibody Clean-Up Kit (Cat. No. 44600) before coupling to magnetic beads
-Make sure the antibody is mixed with the NHS-beads IMMEDIATELY after washing the beads. A delay may cause hydrolysis of the NHS groups on the resin which leads to low coupling efficiency.
-Finally make sure the pH of the non-amine-containing coupling buffer used for coupling is between pH 7-9

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

인용 및 참조 문헌 (4)

인용 및 참조 문헌
Abstract
Peptide barcoding for establishment of new types of genotype-phenotype linkages
Authors:Miyamoto K, Aoki W, Ohtani Y, Miura N, Aburaya S, Matsuzake Y, Kajiwara K, Kitagawa Y, Ueda M
Journal:PLoS ONE
PubMed ID:
Mapping Neutralizing and Immunodominant Sites on the SARS-CoV-2 Spike Receptor-Binding Domain by Structure-Guided High-Resolution Serology
Authors:Piccoli et al.
Journal:Cell
PubMed ID:
ER-luminal [Ca<sup>2+</sup>] regulation of InsP3 receptor gating mediated by an ER-lumianl peripheral Ca<sup>2+</sup> -binding protein
Authors:Vais H, Wang M, Mallilankaraman K, Payne R, McKennan C, Lock JT, Spruce LA, Fiest C, Chan MY, Parker I, Seeholzer SH, Foskett JK, Mak DD
Journal:eLife
PubMed ID:
A single mRNA immunization boosts cross-variant neutralizing antibodies elicited by SARS-CoV-2 infection
Authors:Leonidas Stamatatos, Julie Czartoski, Yu-Hsin Wan, Leah J. Homad, Vanessa Rubin, Hayley Glantz, Moni Neradilek, Emilie Seydoux, Madeleine F. Jennewein, Anna J. MacCamy, Junli Feng, Gregory Mize, Stephen C. De Rosa, Andr&eacute;s Finzi, Maria P. Lemos, Kristen W. Cohen, Zoe Moodie, M. Juliana McElrath, Andrew T. McGuire
Journal:Science
PubMed ID: