Isotemp™ Digital Dry Baths/Block Heaters
Fisher Scientific™

Isotemp™ Digital Dry Baths/Block Heaters

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of PlacesPlug Type
888600234US
888600211US
888600222US
Catalog number 88860023
Price (MXN)
-
No. of Places:
4
Plug Type:
US
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Specifications
Certifications/ComplianceCE, cULus, RoHS
Controller TypePID
Dimensions (L x W x H)17.7 x 7.9 x 3.9 in. (450 x 200 x 100 mm)
Electrical Requirements100/120 V, 50/60 Hz
Frequency50/60 Hz
Fuse250V 2.5A
Heat Up Time≤ 20 min. 30° to 130°C
Height (English)3.9 in.
Height (Metric)100 mm
IncludesDry bath, US style plug, block lifter
Length (English)17.7 in.
Length (Metric)450 mm
Plug TypeUS
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 1°C
Timer0 to 99:59 min or continuous
TypeDry Bath Block
Warranty2 Year, Replacement
Weight (English)10.36 lb.
Weight (Metric)4.7 kg
Width (English)7.9 in.
Width (Metric)200 mm
MaterialPowder-coated Steel
No. of Places4
Relative Humidity<80%
Temperature Range (Metric)Ambient +5° to 130°C
Voltage100/120 V
Unit SizeEach