Thermo Scientific™
Compact Digital Rocker
Related applications:
Lab Equipment
Ensure gentle sample agitation with the Thermo Scientific™ Compact Digital Rocker—a compact, adjustable angle unit.
Back to top
Compact design ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60°C). PID control ensures consistent and smooth rocking motion.
  • Digital Speed Control: 5 to 100rpm
  • Easy to adjust tilt angle: 7° to 13°
  • Timer Range: Continuous or 1 min. to 99 hr., 59 min.
Includes:
Dimpled mat (tray, silicone pad, two securing bands).
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.