Accessories for Tube Revolver
Accessories for Tube Revolver
Actual product may vary
Thermo Scientific™

Accessories for Tube Revolver

Optimize the performance of your Thermo Scientific™ Tube Revolver by using these accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
88881007Paddles 1.5 mL/2mL
88881005Paddles 10mL/15 mL
88881008Paddles 0.5mL/0.8mL
88881006Paddles 5 mL/7mL
88881009Paddles 50mL
Catalog number 88881007
Price (USD)
27.10
Pack of 2
Add to cart
Description:
Paddles 1.5 mL/2mL
Price (USD)
27.10
Pack of 2
Add to cart
Optimize performance by using Thermo Scientific™ Accessories for Tube Revolver.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionPaddles 1.5 mL/2mL
For Use With (Equipment)Rotator/Rocker
HoldsTubes
Product TypeRotisserie Paddle
Tube Capacity1.5/2 mL
Unit SizePack of 2