Accessories for Digital Rockers and Waving Rotators
Thermo Scientific™
Accessories for Digital Rockers and Waving Rotators
Get assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
88882102Dimpled Rubber Mat
88882101Rubber Mat
88882103Rubber Strips, 6 Each
88882128Rubber tube replacement
Catalog number 88882102
Price (USD)
47.18
Online exclusive
55.50
Save 8.32
Pack of 1
Description:
Dimpled Rubber Mat
Price (USD)
47.18
Online exclusive
55.50
Save 8.32
Pack of 1
Assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators
Specifications
DescriptionDimpled Rubber Mat
Dimensions (L x W)298 x 258 mm
For Use With (Equipment)Digital Rockers, Digital Waving Rotator
MaterialRubber
Product TypeDimpled Rubber Mat
Unit SizePack of 1