Accessories for Digital Vortex Mixers
Thermo Scientific™

Accessories for Digital Vortex Mixers

Get assorted accessories for Digital Vortex Mixers
製品番号(カタログ番号)概要寸法(長さ☓幅)材料
8888212319 x 10 mm dia.Tube Holder87.5 x 40 mmFoam, plastic
88882120Rubber Platform82 x 16.5 mmPlastic
88882121Flat bottom vessel platform87.5 x 10 mmRubber
88882122Microplate Tray136.1 x 97.3 mmPlastic
8888212413 x 12 mm dia. Tube Holder87.5 x 40 mmFoam, plastic
8888212512 x 15 mm dia. Tube Holder87.5 x 55 mmFoam, plastic
888821267 x 20 mm dia. Tube Holder87.5 x 55 mmFoam, plastic
888821274 x 26 mm dia.Tube Holder87.5 x 55 mmFoam, plastic
製品番号(カタログ番号) 88882123
価格(JPY)
-
概要:
19 x 10 mm dia.Tube Holder
寸法(長さ☓幅):
87.5 x 40 mm
材料:
Foam, plastic
Assorted accessories for Digital Vortex Mixers
仕様
Black
概要19 x 10 mm dia.Tube Holder
寸法(長さ☓幅)87.5 x 40 mm
使用対象 (装置)Digital Vortex mixer
材料Foam, plastic
製品タイプTube Holder
チューブサイズ10 mm dia.
Unit SizePack of 1