Thermo Scientific™
Richard-Allan Scientific™ May-Grunwald Stain, bottle
Related applications:
Anatomical Pathology
Produce very dense color saturation with Thermo Scientific™ Richard-Allan Scientific™ May-Grunwald Stain, a classical hematology stain
Back to top
  • Requires longer staining time
  • Dramatically darker than Giemsa and Wright Giemsa stains
  • Used in the staining of peripheral blood smears and bone marrows