I-PER™ Insect Cell Protein Extraction Reagent
Thermo Scientific™

I-PER™ Insect Cell Protein Extraction Reagent

Thermo Scientific I-PER Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculovirus-infected insect cellsRead more
Have Questions?
Catalog NumberQuantity
89802250 mL
Catalog number 89802
Price (USD)
330.65
Online exclusive
355.00
Save 24.35
Each
Quantity:
250 mL
Price (USD)
330.65
Online exclusive
355.00
Save 24.35
Each
Thermo Scientific I-PER Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculovirus-infected insect cells grown in suspension or monolayer culture.

Features of I-PER Insect Cell Protein Extraction Reagent :

• Gentle extraction—optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells
• Effective—provides better protein extraction than sonication
• Compatible—downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography
• Flexible—useful for protein extraction from suspended or adherent cultured insect cells

The baculovirus insect cell expression system is an efficient and popular system for the production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
Quantity250 mL
Volume (Metric)250 mL
Product LineI-PER
Product TypeReagent
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for the I-PER Insect Cell Protein Extraction Reagent?

The I-PER Insect Cell Protein Extraction Reagent is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (2)

Citations & References
Abstract
Rift Valley fever virus incorporates the 78 kDa glycoprotein into virions matured in mosquito C6/36 cells.
Authors:Weingartl HM, Zhang S, Marszal P, McGreevy A, Burton L, Wilson WC
Journal:
PubMed ID:24489907
'Rift Valley fever virus (RVFV), genus Phlebovirus, family Bunyaviridae is a zoonotic arthropod-borne virus able to transition between distant host species, causing potentially severe disease in humans and ruminants. Viral proteins are encoded by three genomic segments, with the medium M segment coding for four proteins: nonstructural NSm protein, two ... More
Tnao38, high five and Sf9--evaluation of host-virus interactions in three different insect cell lines: baculovirus production and recombinant protein expression.
Authors:Wilde M, Klausberger M, Palmberger D, Ernst W, Grabherr R
Journal:
PubMed ID:24375231
A comparative analysis of new and established insect cell lines, in regard to process relevant parameters, provide data that can be exploited for designing more robust and effective protein production processes. The baculovirus-insect cell expression system has been efficiently used for the production of heterologous proteins. Three different insect cell ... More