Pierce™ C18 Spin Columns

Catalog number: 89870B

Thermo Scientific™  Related applications: Protein Mass Spectrometry Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce C18 Spin Columns are ready-to-use centrifuge columns of porous C18 reverse-phase resin with excellent binding and recovery characteristics for peptide sample preparation for mass spectrometry and other methods.

Peptide samples can be purified and concentrated for a variety of applications using Pierce C18 Spin Columns. Each spin column contains a porous C18 reversed-phase resin with excellent binding and recovery characteristics for a wide range of peptide concentrations. The spin column format allows simultaneous processing of multiple samples (10 to 150 µL each) in approximately 30 minutes without laborious repeat pipetting or specialized equipment. Pierce C18 Spin Columns can be used effectively for processing peptides derived from 10 ng to 30 µg of protein. Sensitivity and detection limits are dependent on the downstream application.

Features of C18 Spin Columns:

Removes interfering contaminants—significantly reduces signal suppression and improves signal-to-noise ratios and sequence coverage
Simplifies optimization—processing yields high-quality spectra and is effective for a variety of reverse-phase-compatible contaminates
Robust—works with a wide variety of load volumes and concentrations; no need to reduce sample volume before application
Convenient—easy to handle and requires no special equipment to process multiple samples simultaneously (unlike tip-driven systems that require one sample to be processed at a time)
Sensitive—special C18 resin allows excellent recovery percentages, even at low (sub-picomole) sample loads

Matrix-assisted laser desorption ionization (MALDI) and electrospray ionization (ESI) mass spectrometry (MS) are essential for studying biological compounds because of the high sensitivity and mass accuracy they provide. However, many of the buffers and compounds common to biological samples interfere with both MALDI-MS and ESI-MS, resulting in reduced sensitivity and poor-quality spectra. Pierce C-18 Spin Columns remove interfering contaminants and release peptides in MS-compatible solutions, providing an effective tool that aids in the MS examination of post-translational modifications or for the identification of proteins by peptide mapping.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Mass Spectrometry
Final Product: Peptide
For Use With (Equipment): Mass Spectrometer, Microcentrifuge
Product Line: Pierce™
Product Size: Custom
Starting Material: Peptides, Protease-digested protein
Workflow Step: Sample Clean Up

Contents & storage

Store at room temperature.

Documents

Manuals & protocols