EDTA disodium salt dihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

EDTA disodium salt dihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8995-01500 g
8995-20100 lb.
8995-0712 kg
Catalog number 8995-01
Price (MXN)
-
Quantity:
500 g

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.

Specifications
CAS6381-92-6
Chemical Name or MaterialEDTA disodium salt dihydrate
ColorWhite
Melting Point255°C
Molecular FormulaC10H14N2Na2O8·2H2O
Packaging TypeAmber glass bottle, wide mouth, tamper-evident cap
Purity99.0 to 101.0%
Quantity500 g
SynonymEthylenediaminetetra-acetic acid disodium salt dihydrate
GradeMulti-Compendial/NF
Unit SizeEach